missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxycinnamic acid, 98+%

$78.04 - $292.46

Chemical Identifiers

CAS 69038-81-9
Molecular Formula C11H12O3
Formula Weight 192.22
MDL Number MFCD00016839
Beilstein 2693328
Chemical Name or Material 2-Ethoxycinnamic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0665006 SDS Alfa Aesar™
L0665006
5g Each for $78.04
AAL0665014 SDS Alfa Aesar™
L0665014
25g Each for $292.46
Specifications Description & Specifications

Chemical Identifiers

69038-81-9
192.22
2693328
C11H12O3
MFCD00016839
2-Ethoxycinnamic acid

Specifications

134° to 136°C
69038-81-9
192.22
2693328
≥98%
5g
C11H12O3
MFCD00016839
2-Ethoxycinnamic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning