missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethoxycarbonyl)phenyl isothiocyanate, 98+%

Manufacturer:  Alfa Aesar™ L1256906

 View more versions of this product

Catalog No. AAL1256906



Specifications

Specifications

2-(Ethoxycarbonyl)phenyl isothiocyanate
144°C to 145°C (8mmHg)
99960-09-5
MFCD00041063
3267738
TYEPWLJHQLYWSR-UHFFFAOYSA-N
ethyl 2-isothiocyanatobenzoate
523235
≥98%
>110°C (230°F)
5g
C10H9NO2S
UN2922
2-ethoxycarbonylphenyl isothiocyanate, 2-ethoxycarbonyl phenyl isothiocyanate, 2-isothiocyanatobenzoic acid ethyl ester, 2-isothiocyanato-benzoic acid ethyl ester, acmc-20aon5, #, 2-ethoxycarbonylphenylisothiocyanate, ethyl 2-isothiocyanatobenzoate, isothiocyanatobenzoic acid ethyl ester, benzoic acid,2-isothiocyanato-,ethyl ester
CCOC(=O)C1=CC=CC=C1N=C=S
207.247
207.25

Chemical Identifiers

99960-09-5
207.247
TYEPWLJHQLYWSR-UHFFFAOYSA-N
523235
CCOC(=O)C1=CC=CC=C1N=C=S
C10H9NO2S
MFCD00041063
2-ethoxycarbonylphenyl isothiocyanate, 2-ethoxycarbonyl phenyl isothiocyanate, 2-isothiocyanatobenzoic acid ethyl ester, 2-isothiocyanato-benzoic acid ethyl ester, acmc-20aon5, #, 2-ethoxycarbonylphenylisothiocyanate, ethyl 2-isothiocyanatobenzoate, isothiocyanatobenzoic acid ethyl ester, benzoic acid,2-isothiocyanato-,ethyl ester
ethyl 2-isothiocyanatobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents