Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethoxycarbonyl)phenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 2-(Ethoxycarbonyl)phenyl isocyanate
CAS 76393-16-3
Molecular Formula C10H9NO3
Formula Weight 191.19g/mol
MDL Number MFCD00013849
UN Number UN2811
Beilstein 1242648
Synonym 2-Carboethoxyphenyl isocyanate; Ethyl 2-isocyanatobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0956003

 
alfa aesar™
L09560-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 27° to 31°C
Quantity 1g
Flash Point >110°C (230°F)
Chemical Name or Material 2-(Ethoxycarbonyl)phenyl isocyanate
CAS 76393-16-3
Assay 98%
Molecular Formula C10H9NO3
Formula Weight 191.19g/mol
MDL Number MFCD00013849
UN Number UN2811
Beilstein 1242648
Synonym 2-Carboethoxyphenyl isocyanate; Ethyl 2-isocyanatobenzoate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C