Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethoxycarbonyl)ethylzinc bromide, 0.5M in THF

Chemical Identifiers

Chemical Name or Material 2-(Ethoxycarbonyl)ethylzinc bromide
CAS 193065-68-8
Molecular Formula BrZnCH2CH2COOCH2CH3
Formula Weight 246.42
MDL Number MFCD00671976
UN Number UN3399
Synonym 3-Ethoxy-3-oxopropylzinc bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26739AD

  SDS
alfa aesar™
H26739AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.42
Health Hazard 1 Danger
Chemical Name or Material 2-(Ethoxycarbonyl)ethylzinc bromide
Concentration 0.5M in THF
Molecular Formula BrZnCH2CH2COOCH2CH3
MDL Number MFCD00671976
CAS 193065-68-8
UN Number UN3399
Synonym 3-Ethoxy-3-oxopropylzinc bromide
Quantity 50mL
Flash Point −17°C (1°F)
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

Applications
In many of the reactions, organozinc compounds appear as intermediates. Some of the synthetic uses of organozinc compounds are: for the stereo selective alkylation reaction of cyclic ketones, conjugate addition reaction to alpha, beta-unsaturated carbonyl compounds, ring opening reactions of epoxides and oxo vanadium-induced ligand coupling of aryltrimethyl zincates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture and heat.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C