missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid, 95%, Alfa Aesar™

$31.61 - $370.11

Chemical Identifiers

CAS 133944-74-8
Molecular Formula C7H12O3S2
Molecular Weight (g/mol) 208.29
MDL Number MFCD19689494
InChI Key FBDXTAKUEUFSGH-UHFFFAOYSA-N
Synonym 2-ethoxythioxomethyl thio-2-methylpropanoic acid, 2-ethoxythioxo methyl thio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoic acid, propanoic acid, 2-ethoxythioxomethyl thio-2-methyl, 2-ethoxymethanethioyl sulfanyl-2-methylpropanoic acid, ksc493s4h, 2-ethoythioxo methyl thio-isobutyric acid, alpha-ethoxythiocarbonylmercapto-isobutyric acid, 2-ethoxycarbonothioylthio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoicacid
PubChem CID 53249283
IUPAC Name 2-ethoxycarbothioylsulfanyl-2-methylpropanoic acid
SMILES CCOC(=S)SC(C)(C)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66036MD View Documents Alfa Aesar
H66036MD
250mg Each for $31.61
 
AAH6603603 View Documents Alfa Aesar
H6603603
1g Each for $93.30
 
AAH6603606 View Documents Alfa Aesar
H6603606
5g Each for $370.11
 
Specifications

Chemical Identifiers

133944-74-8
208.29
FBDXTAKUEUFSGH-UHFFFAOYSA-N
53249283
CCOC(=S)SC(C)(C)C(=O)O
C7H12O3S2
MFCD19689494
2-ethoxythioxomethyl thio-2-methylpropanoic acid, 2-ethoxythioxo methyl thio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoic acid, propanoic acid, 2-ethoxythioxomethyl thio-2-methyl, 2-ethoxymethanethioyl sulfanyl-2-methylpropanoic acid, ksc493s4h, 2-ethoythioxo methyl thio-isobutyric acid, alpha-ethoxythiocarbonylmercapto-isobutyric acid, 2-ethoxycarbonothioylthio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoicacid
2-ethoxycarbothioylsulfanyl-2-methylpropanoic acid

Specifications

250mg
C7H12O3S2
1779889
FBDXTAKUEUFSGH-UHFFFAOYSA-N
2-ethoxycarbothioylsulfanyl-2-methylpropanoic acid
53249283
Solid
2-[(Ethoxycarbonothioyl)thio]-2-methylpropionic acid
133944-74-8
MFCD19689494
2-ethoxythioxomethyl thio-2-methylpropanoic acid, 2-ethoxythioxo methyl thio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoic acid, propanoic acid, 2-ethoxythioxomethyl thio-2-methyl, 2-ethoxymethanethioyl sulfanyl-2-methylpropanoic acid, ksc493s4h, 2-ethoythioxo methyl thio-isobutyric acid, alpha-ethoxythiocarbonylmercapto-isobutyric acid, 2-ethoxycarbonothioylthio-2-methylpropanoic acid, 2-ethoxythioxomethyl thio-2-methylpropanoicacid
CCOC(=S)SC(C)(C)C(=O)O
208.29
208.29
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning