missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2-Ethoxybenzyl alcohol, 98%
Manufacturer: Alfa Aesar™ A1547714
Description
Applications2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product. It is also used as pharmaceutical intermediates.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
2-Ethoxybenzyl alcohol | |
263°C to 265°C | |
71672-75-8 | |
MFCD00004616 | |
2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn | |
CCOC1=CC=CC=C1CO | |
152.193 | |
152.19 |
>110°C (230°F) | |
25g | |
C9H12O2 | |
2555374 | |
ICJVQAHPHKYCNU-UHFFFAOYSA-N | |
(2-ethoxyphenyl)methanol | |
51381 | |
98% |
Chemical Identifiers
71672-75-8 | |
152.193 | |
ICJVQAHPHKYCNU-UHFFFAOYSA-N | |
51381 | |
CCOC1=CC=CC=C1CO |
C9H12O2 | |
MFCD00004616 | |
2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn | |
(2-ethoxyphenyl)methanol |
Safety and Handling
RTECSNumber : DO5467000