missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxybenzoyl chloride, 98%

$154.22 - $154.22

Chemical Identifiers

CAS 42926-52-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00016348
InChI Key MDKAAWDKKBFSTK-UHFFFAOYSA-N
Synonym o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride
PubChem CID 3016397
IUPAC Name 2-ethoxybenzoyl chloride
SMILES CCOC1=CC=CC=C1C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1768622 View Documents Alfa Aesar™
L1768622
100g Each for $154.22
 
Specifications

Chemical Identifiers

42926-52-3
184.619
MDKAAWDKKBFSTK-UHFFFAOYSA-N
3016397
CCOC1=CC=CC=C1C(=O)Cl
C9H9ClO2
MFCD00016348
o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride
2-ethoxybenzoyl chloride

Specifications

114°C to 118°C (5mmHg)
42926-52-3
MFCD00016348
o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride
CCOC1=CC=CC=C1C(=O)Cl
184.619
184.62
2-Ethoxybenzoyl chloride
100g
C9H9ClO2
UN3265
MDKAAWDKKBFSTK-UHFFFAOYSA-N
2-ethoxybenzoyl chloride
3016397
98%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.