missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxybenzonitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435340050

 View more versions of this product

Catalog No. AC435340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Ethoxybenzonitrile
Authentic
Colorless to Yellow
5g
97.5
97.5% min. (GC)
MFCD00001789
DXTLCLWOCYLDHL-UHFFFAOYSA-N
2-ethoxybenzonitrile
576922
Liquid
113°C
45°C (0.5 Torr)
5°C
6609-57-0
100.0
C9H9NO
2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, 2-ethoxybenzenecarbonitrile, 2-ethoxybenzonitrile, acmc-20anwu, benzonitrile, 2-ethoxy, benzonitrile, ethoxy, o-ethoxybenzonitrile, o-ethoxybenzontrle, pubchem4773
CCOC1=CC=CC=C1C#N
147.18
147.18
98%

Chemical Identifiers

6609-57-0
147.18
DXTLCLWOCYLDHL-UHFFFAOYSA-N
576922
CCOC1=CC=CC=C1C#N
C9H9NO
MFCD00001789
2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, 2-ethoxybenzenecarbonitrile, 2-ethoxybenzonitrile, acmc-20anwu, benzonitrile, 2-ethoxy, benzonitrile, ethoxy, o-ethoxybenzonitrile, o-ethoxybenzontrle, pubchem4773
2-ethoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

SDS Documents