Please login to your online account to display your discounted pricing

2-Ethoxybenzonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Ethoxybenzonitrile
CAS 6609-57-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00001789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435340010

  SDS
thermo fisher scientific
AC435340010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC435340050

  SDS
thermo fisher scientific
AC435340050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 147.18
Quantity 1g
Color Clear colorless to pale yellow
CAS Max % 100.0
MDL Number MFCD00001789
CAS 6609-57-0
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Ethoxybenzonitrile
Assay Percent Range 98%
Molecular Formula C9H9NO
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.