Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxybenzoic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Ethoxybenzoic acid
CAS 134-11-2
Formula Weight 166.18
MDL Number MFCD00002438
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1546818

 
alfa aesar™
1546818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1546830

 
alfa aesar™
1546830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material 2-Ethoxybenzoic acid
TSCA TSCA
Physical Form Liquid
Assay 98%
MDL Number MFCD00002438
CAS 134-11-2
Refractive Index 1.5400
Density 1.105g/mL
Boiling Point 174° to 176°C (15mmHg)
Melting Point 19°C
Quantity 50g
Flash Point 113° (235°F)