Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxybenzeneboronic acid, 98%

$52.12 - $596.68

Chemical Identifiers

Chemical Name or Material 2-Ethoxybenzeneboronic acid
CAS 213211-69-9
Molecular Formula C8H11BO3
Formula Weight 165.99
MDL Number MFCD00674027
Synonym 2-Ethoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2364403

  SDS
alfa aesar™
B2364403
Each for $52.12

AAB2364414

  SDS
alfa aesar™
B2364414
Each for $596.68
Description & Specifications

Specifications

Formula Weight 165.99
MDL Number MFCD00674027
Melting Point 98° to 101°C
Molecular Formula C8H11BO3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Chemical Name or Material 2-Ethoxybenzeneboronic acid
CAS 213211-69-9
Synonym 2-Ethoxyphenylboronic acid
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.