missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxybenzaldehyde, 97+%

Manufacturer:  Alfa Aesar™ A1503530

 View more versions of this product

Catalog No. AAA1503530



Specifications

Specifications

2-Ethoxybenzaldehyde
249°C
613-69-4
MFCD00003316
benzaldehyde, 2-ethoxy, o-ethoxybenzaldehyde, 2-ethoxy benzaldehyde, benzaldehyde, o-ethoxy, 2-ethoxy-benzaldehyde, benzaldehyde, ethoxy, ethoxy benzaldehyde, pubchem9726, ortho-ethoxybenzaldehyde, acmc-209ms1
CCOC1=CC=CC=C1C=O
150.177
150.18
91°C (195°F)
250g
C9H10O2
1937270
DUVJMSPTZMCSTQ-UHFFFAOYSA-N
2-ethoxybenzaldehyde
11950
≥97%

Chemical Identifiers

613-69-4
150.177
DUVJMSPTZMCSTQ-UHFFFAOYSA-N
11950
CCOC1=CC=CC=C1C=O
C9H10O2
MFCD00003316
benzaldehyde, 2-ethoxy, o-ethoxybenzaldehyde, 2-ethoxy benzaldehyde, benzaldehyde, o-ethoxy, 2-ethoxy-benzaldehyde, benzaldehyde, ethoxy, ethoxy benzaldehyde, pubchem9726, ortho-ethoxybenzaldehyde, acmc-209ms1
2-ethoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents