Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Ethoxyacetophenone, 97+%

$81.22 - $310.39

Chemical Identifiers

Chemical Name or Material 2'-Ethoxyacetophenone
CAS 2142-67-8
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00017262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011309

  SDS
alfa aesar™
B2011309
10g Each for $81.22

AAB2011318

  SDS
alfa aesar™
B2011318
50g Each for $310.39
Description & Specifications

Specifications

Formula Weight 164.2
Quantity 10g
CAS 2142-67-8
Assay 98%
MDL Number MFCD00017262
Melting Point 38° to 42°C
Boiling Point 244°C
Chemical Name or Material 2'-Ethoxyacetophenone
Flash Point >110°C (230°F)
Molecular Formula C10H12O2