missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethoxy-5-nitropyridine 98.0+%, TCI America™
$99.54 - $308.57
Chemical Identifiers
CAS | 31594-45-3 |
---|---|
Molecular Formula | C7H8N2O3 |
Molecular Weight (g/mol) | 168.15 |
MDL Number | MFCD01646181 |
InChI Key | LYDVDLYMQVSLQD-UHFFFAOYSA-N |
PubChem CID | 295864 |
IUPAC Name | 2-ethoxy-5-nitropyridine |
SMILES | CCOC1=CC=C(C=N1)[N+]([O-])=O |
Chemical Identifiers
31594-45-3 | |
168.15 | |
LYDVDLYMQVSLQD-UHFFFAOYSA-N | |
2-ethoxy-5-nitropyridine |
C7H8N2O3 | |
MFCD01646181 | |
295864 | |
CCOC1=CC=C(C=N1)[N+]([O-])=O |
Specifications
31594-45-3 | |
C7H8N2O3 | |
5 g | |
CCOC1=CC=C(C=N1)[N+]([O-])=O | |
168.15 | |
168.15 | |
Crystalline Powder |
Yellow | |
MFCD01646181 | |
LYDVDLYMQVSLQD-UHFFFAOYSA-N | |
2-ethoxy-5-nitropyridine | |
295864 | |
≥98.0% (GC) | |
2-Ethoxy-5-nitropyridine |
Safety and Handling
TSCA : No