missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxy-5-methylbenzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H5334003

 View more versions of this product

Catalog No. AAH5334003



Specifications

Specifications

2-Ethoxy-5-methylbenzeneboronic acid
1g
C9H13BO3
2-ethoxy-5-methylbenzeneboronic acid, 2-ethoxy-5-methylphenyl boronic acid, 2-ethoxy-5-methyl-phenyl boronic acid, 2-ethoxy-5-methylphenylboronicacid, 4-ethoxytoluene-3-boronic acid, acmc-1c5pf, contains varying amounts of anhydride
B(C1=C(C=CC(=C1)C)OCC)(O)O
180.01
180.01
98°C to 101°C
123291-97-4
MFCD00196832
KBGGTPGSWHNQEX-UHFFFAOYSA-N
(2-ethoxy-5-methylphenyl)boronic acid
4615191
95%

Chemical Identifiers

123291-97-4
180.01
KBGGTPGSWHNQEX-UHFFFAOYSA-N
4615191
B(C1=C(C=CC(=C1)C)OCC)(O)O
C9H13BO3
MFCD00196832
2-ethoxy-5-methylbenzeneboronic acid, 2-ethoxy-5-methylphenyl boronic acid, 2-ethoxy-5-methyl-phenyl boronic acid, 2-ethoxy-5-methylphenylboronicacid, 4-ethoxytoluene-3-boronic acid, acmc-1c5pf, contains varying amounts of anhydride
(2-ethoxy-5-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents