missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxy-5-fluoro-4-pyrimidinone, TCI America™

Manufacturer:  TCI America E068425G

 View more versions of this product

Catalog No. E068425G



Specifications

Specifications

2-Ethoxy-5-fluoro-4-pyrimidinone
180°C
56177-80-1
5-Fluoro-2-ethoxy-4-pyrimidinone
CCOC1=NC=C(C(=O)N1)F
158.132
158.13
≥98.0% (GC)
White
25g
C6H7FN2O2
XREDGJFKPWBNRQ-UHFFFAOYSA-N
2-ethoxy-5-fluoro-1H-pyrimidin-6-one
92041
Crystal-Powder at 20°C

Chemical Identifiers

56177-80-1
158.132
5-Fluoro-2-ethoxy-4-pyrimidinone
2-ethoxy-5-fluoro-1H-pyrimidin-6-one
C6H7FN2O2
XREDGJFKPWBNRQ-UHFFFAOYSA-N
92041
CCOC1=NC=C(C(=O)N1)F
Safety and Handling

Safety and Handling

TSCA : No

Documents