Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxy-4-nitrobenzoic acid, 98%

$84.82 - $272.73

Chemical Identifiers

Chemical Name or Material 2-Ethoxy-4-nitrobenzoic acid
CAS 2486-66-0
Molecular Formula C9H9NO5
Formula Weight 211.18
MDL Number MFCD02683865
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2721706

  SDS
alfa aesar™
H2721706
5g Each for $84.82

AAH2721714

  SDS
alfa aesar™
H2721714
25g Each for $272.73
Description & Specifications

Specifications

Formula Weight 211.18
Chemical Name or Material 2-Ethoxy-4-nitrobenzoic acid
Assay 98%
Molecular Formula C9H9NO5
MDL Number MFCD02683865
CAS 2486-66-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 146° to 151°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.