missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxy-4-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H55874MD

 View more versions of this product

Catalog No. AAH55874MD



Specifications

Specifications

2-Ethoxy-4-fluorobenzeneboronic acid
250mg
C8H10BFO3
2-ethoxy-4-fluoranyl-phenyl boronic acid
B(C1=C(C=C(C=C1)F)OCC)(O)O
183.973
183.97
136°C to 141°C
480438-58-2
MFCD03095001
WCJKVQYVILAKQW-UHFFFAOYSA-N
(2-ethoxy-4-fluorophenyl)boronic acid
16217201
97%

Chemical Identifiers

480438-58-2
183.973
WCJKVQYVILAKQW-UHFFFAOYSA-N
16217201
B(C1=C(C=C(C=C1)F)OCC)(O)O
C8H10BFO3
MFCD03095001
2-ethoxy-4-fluoranyl-phenyl boronic acid
(2-ethoxy-4-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents