missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxy-3-methylpyrazine, TCI America™

Manufacturer:  TCI America M080925G

 View more versions of this product

Catalog No. M080925G



Specifications

Specifications

2-Ethoxy-3-methylpyrazine
Colorless
32737-14-7
MFCD00038025
CCOC1=NC=CN=C1C
138.17
138.17
≥98.0% (GC,T)
88°C
25g
C7H10N2O
MMKWCKGYULOKET-UHFFFAOYSA-N
2-ethoxy-3-methylpyrazine
122941
Clear Liquid at 20°C

Chemical Identifiers

32737-14-7
138.17
MMKWCKGYULOKET-UHFFFAOYSA-N
2-ethoxy-3-methylpyrazine
C7H10N2O
MFCD00038025
122941
CCOC1=NC=CN=C1C
Safety and Handling

Safety and Handling

TSCA : Yes

Documents