Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxy-1-naphthoic acid, 98%

$261.68 - $261.68

Chemical Identifiers

Chemical Name or Material 2-Ethoxy-1-naphthoic acid
CAS 2224-00-2
Molecular Formula C13H12O3
Formula Weight 216.24
MDL Number MFCD00004008
Beilstein 2695612
Synonym 2-Ethoxynaphthalene-1-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2266522

 
alfa aesar™
B2266522
100g Each for $261.68
Description & Specifications

Specifications

Formula Weight 216.24
Chemical Name or Material 2-Ethoxy-1-naphthoic acid
TSCA TSCA
Assay 98%
Molecular Formula C13H12O3
MDL Number MFCD00004008
CAS 2224-00-2
Beilstein 2695612
Synonym 2-Ethoxynaphthalene-1-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 143° to 145°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.