Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxy-1-naphthaldehyde, 98%

$59.38 - $170.27

Chemical Identifiers

Chemical Name or Material 2-Ethoxy-1-naphthaldehyde
CAS 19523-57-0
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD00046427
Beilstein 1073578
Synonym 2-Ethoxynaphthalene-1-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2530603

  SDS
alfa aesar™
B2530603
1g Each for $59.38

AAB2530606

  SDS
alfa aesar™
B2530606
5g Each for $170.27
Description & Specifications

Specifications

Formula Weight 200.24
Chemical Name or Material 2-Ethoxy-1-naphthaldehyde
Assay 98%
Molecular Formula C13H12O2
MDL Number MFCD00046427
CAS 19523-57-0
Beilstein 1073578
Synonym 2-Ethoxynaphthalene-1-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Melting Point 111° to 113°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.