Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dodecanone, 98+%

$124.05 - $124.05

Chemical Identifiers

Chemical Name or Material 2-Dodecanone
CAS 6175-49-1
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00015064
Beilstein 1703135
Synonym n-Decyl methyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1886218

 
alfa aesar™
A1886218
50g Each for $124.05
Description & Specifications

Specifications

Formula Weight 184.32
Chemical Name or Material 2-Dodecanone
TSCA TSCA
Assay ≥98%
Molecular Formula C12H24O
MDL Number MFCD00015064
CAS 6175-49-1
Beilstein 1703135
Synonym n-Decyl methyl ketone
Boiling Point 247°C
Melting Point 17° to 20°C
Flash Point 110°C (230°F)
Quantity 50g