missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Dodecanone, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 330291000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Dodecanone | |
94.0 | |
0.8200g/mL | |
108°C | |
94% min. (GC) | |
C12H24O | |
MFCD00015064 | |
0.82 | |
LSKONYYRONEBKA-UHFFFAOYSA-N | |
dodecan-2-one | |
22556 | |
95% |
6175-49-1 | |
100.0 | |
247°C | |
Authentic | |
Glass bottle | |
1.4320 to 1.4340 (20°C, 589nm) | |
100mL | |
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc | |
CCCCCCCCCCC(=O)C | |
184.32 | |
184.32 |
Chemical Identifiers
6175-49-1 | |
184.32 | |
LSKONYYRONEBKA-UHFFFAOYSA-N | |
22556 | |
CCCCCCCCCCC(=O)C |
C12H24O | |
MFCD00015064 | |
2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc | |
dodecan-2-one |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
Collect spillage.
EINECSNumber : 228-222-5
RUO â Research Use Only