Please login to your online account to display your discounted pricing

2-Dodecanone, 95%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  330290250

 View more versions of this product

Catalog No. AC330290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 184.32
Physical Form Liquid
Quantity 25mL
Color Clear colorless to light yellow
Density 0.8200g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Infrared Spectrum Authentic
Molecular Formula C12H24O
CAS 6175-49-1
Chemical Name or Material 2-Dodecanone
MDL Number MFCD00015064
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)

Specifications

Chemical Name or Material 2-Dodecanone
CAS 6175-49-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00015064