missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Diphenylphosphino)ethyltriethoxysilane 92%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC391860050

 View more versions of this product

Catalog No. AC391860050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-(Diphenylphosphino)ethyltriethoxysilane
Clear colorless to light yellow
18586-39-5
100.0
Glass bottle
Liquid
92%
376.51
1.05g/mL
5g
90.0
Authentic
1.5390 to 1.5410 (20°C, 589nm)
≥90%
C20H29O3PSi
MFCD00015125

Chemical Identifiers

18586-39-5
100.0
C20H29O3PSi
MFCD00015125
≥90%
90.0
92%
376.51
2-(Diphenylphosphino)ethyltriethoxysilane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.