Please login to your online account to display your discounted pricing

2-(Diphenylphosphino)ethyltriethoxysilane 92%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391860010

 View more versions of this product

Catalog No. AC391860010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥90%
Physical Form Liquid
Formula Weight 376.51
Quantity 1g
Color Clear colorless to light yellow
Density 1.05g/mL
CAS Min % 90.0
CAS Max % 100.0
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Infrared Spectrum Authentic
CAS 18586-39-5
Assay Percent Range 92%
Molecular Formula C20H29O3PSi
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00015125
Packaging Glass bottle
Chemical Name or Material 2-(Diphenylphosphino)ethyltriethoxysilane
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-(Diphenylphosphino)ethyltriethoxysilane
CAS 18586-39-5
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90%
Assay Percent Range 92%
Molecular Formula C20H29O3PSi
Formula Weight 376.51
MDL Number MFCD00015125
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents