Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Diphenylphosphino)ethylamine, 95%

Chemical Identifiers

Chemical Name or Material 2-(Diphenylphosphino)ethylamine
CAS 4848-43-5
Molecular Formula (C6H5)2PCH2CH2NH2
Formula Weight 229.27
MDL Number MFCD00233831
UN Number UN2735
Synonym (2-Aminoethyl)diphenylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2707003

  SDS
alfa aesar™
H2707003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2707009

  SDS
alfa aesar™
H2707009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.27
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 1 Danger
UN Number UN2735
MDL Number MFCD00233831
CAS 4848-43-5
Molecular Formula (C6H5)2PCH2CH2NH2
Chemical Name or Material 2-(Diphenylphosphino)ethylamine
Synonym (2-Aminoethyl)diphenylphosphine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.