Learn More
2-(Diphenylphosphino)biphenyl, 98%, Thermo Scientific Chemicals
$149.03 - $149.03
Chemical Identifiers
CAS | 13885-09-1 |
---|---|
Molecular Formula | C24H19P |
Molecular Weight (g/mol) | 338.39 |
MDL Number | MFCD11559063 |
InChI Key | FNCQSSIMHQVKGF-UHFFFAOYSA-N |
Synonym | 2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl |
PubChem CID | 12976978 |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6164006
|
Thermo Scientific Chemicals
H6164006 |
5 g |
Each for $149.03
|
|
|||||
Description
2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
13885-09-1 | |
338.39 | |
FNCQSSIMHQVKGF-UHFFFAOYSA-N | |
12976978 |
C24H19P | |
MFCD11559063 | |
2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1 |
Specifications
13885-09-1 | |
MFCD11559063 | |
2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl | |
FNCQSSIMHQVKGF-UHFFFAOYSA-N | |
diphenyl-(2-phenylphenyl)phosphane | |
12976978 | |
98% | |
2-(Diphenylphosphino)biphenyl |
C24H19P | |
5 g | |
Slightly soluble in water. | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1 | |
338.39 | |
338.38 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only