Please login to your online account to display your discounted pricing

2-(Diphenylphosphino)benzoic Acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC382930010

 View more versions of this product

Catalog No. AC382930010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 306.3
Quantity 1g
Color Light yellow
Melting Point 177° to 181°C
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C6H5)2PC6H4COOH
MDL Number MFCD00674024
CAS 17261-28-8
Chemical Name or Material 2-(Diphenylphosphino)benzoic acid
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C19H15O2P
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Diphenylphosphino)benzoic acid
CAS 17261-28-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C19H15O2P
Linear Formula (C6H5)2PC6H4COOH
Formula Weight 306.3
MDL Number MFCD00674024
Accessories

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents