Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Diphenylphosphino)benzoic acid, 97%

$68.86 - $926.66

Chemical Identifiers

Chemical Name or Material 2-(Diphenylphosphino)benzoic acid
CAS 17261-28-8
Molecular Formula C19H15O2P
Formula Weight 306.31
MDL Number MFCD00674024
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2238903

  SDS
alfa aesar™
B2238903
1g Each for $68.86

AAB2238906

  SDS
alfa aesar™
B2238906
5g Each for $227.53

AAB2238914

  SDS
alfa aesar™
B2238914
25g Each for $926.66
Description & Specifications

Specifications

Formula Weight 306.31
Assay 97%
Health Hazard 1 Warning
Melting Point 174° to 181°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Health Hazard 2 GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(Diphenylphosphino)benzoic acid
Quantity 1g
MDL Number MFCD00674024
CAS 17261-28-8
RTECS Number DG9287500
Molecular Formula C19H15O2P
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.