missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Diphenylphosphino-2'-methylbiphenyl, 98%

Manufacturer:  Alfa Aesar™ H6168503

 View more versions of this product

Catalog No. AAH6168503



Specifications

Specifications

2-Diphenylphosphino-2'-methylbiphenyl
402822-72-4
MFCD19441232
GUENDLLDUNRPIC-UHFFFAOYSA-N
[2-(2-methylphenyl)phenyl]-diphenylphosphane
15531745
Solid
1g
C25H21P
2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
352.417
352.41
98%

Chemical Identifiers

402822-72-4
352.417
GUENDLLDUNRPIC-UHFFFAOYSA-N
15531745
CC1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
C25H21P
MFCD19441232
2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
[2-(2-methylphenyl)phenyl]-diphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents