missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Diphenylphosphino-2'-methylbiphenyl, 98%

Manufacturer:  Alfa Aesar™ H6168506

 View more versions of this product

Catalog No. AAH6168506



Specifications

Specifications

2-Diphenylphosphino-2'-methylbiphenyl
402822-72-4
MFCD19441232
GUENDLLDUNRPIC-UHFFFAOYSA-N
[2-(2-methylphenyl)phenyl]-diphenylphosphane
15531745
Solid
5g
C25H21P
2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
352.417
352.41
98%

Chemical Identifiers

402822-72-4
352.417
GUENDLLDUNRPIC-UHFFFAOYSA-N
15531745
CC1=CC=CC=C1C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
C25H21P
MFCD19441232
2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
[2-(2-methylphenyl)phenyl]-diphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents