missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335620050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride | |
42036-65-7 | |
100.0 | |
White | |
98% | |
C9H17NO·HCl | |
5 g | |
Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol | |
CN(C)CC1CCCCC1=O.Cl | |
191.7 | |
191.7 | |
Crystalline Powder |
98% | |
97.5 | |
147°C to 151°C | |
Authentic | |
Glass bottle | |
MFCD00102580 | |
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp | |
CLVHTSWMNNSUSH-UHFFFAOYSA-N | |
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride | |
359482 | |
98% |
Chemical Identifiers
42036-65-7 | |
191.7 | |
CLVHTSWMNNSUSH-UHFFFAOYSA-N | |
359482 | |
CN(C)CC1CCCCC1=O.Cl |
C9H17NO·HCl | |
MFCD00102580 | |
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp | |
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only