missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Dimethylaminoethyl)triphenylphosphonium bromide, 98%

$48.58 - $177.34

Chemical Identifiers

CAS 21331-80-6
Molecular Formula C22H25BrNP
Molecular Weight (g/mol) 414.327
MDL Number MFCD00011794
InChI Key YIIHEMGEQQWSMA-UHFFFAOYSA-M
Synonym 2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide, 2-dimethylaminoethyl triphenylphosphonium bromide
PubChem CID 2724136
IUPAC Name 2-(dimethylamino)ethyl-triphenylphosphanium;bromide
SMILES CN(C)CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0091709 View Documents Alfa Aesar™
L0091709
10g Each for $48.58
 
AAL0091718 View Documents Alfa Aesar™
L0091718
50g Each for $177.34
 
Specifications

Chemical Identifiers

21331-80-6
414.327
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2724136
CN(C)CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C22H25BrNP
MFCD00011794
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide, 2-dimethylaminoethyl triphenylphosphonium bromide
2-(dimethylamino)ethyl-triphenylphosphanium;bromide

Specifications

~200°C
21331-80-6
MFCD00011794
2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide, 2-dimethylaminoethyl triphenylphosphonium bromide
CN(C)CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
414.327
414.33
(2-Dimethylaminoethyl)triphenylphosphonium bromide
10g
C22H25BrNP
4072092
YIIHEMGEQQWSMA-UHFFFAOYSA-M
2-(dimethylamino)ethyl-triphenylphosphanium;bromide
2724136
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning