Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dimethylaminoethyl chloride hydrochloride, 98+%

Chemical Identifiers

Chemical Name or Material 2-Dimethylaminoethyl chloride hydrochloride
CAS 4584-46-7
Molecular Formula ClCH2CH2N(CH3)2·HCl
Formula Weight 144.05
MDL Number MFCD00012516
UN Number UN2811
Beilstein 3591234
Synonym 1-Chloro-2-(dimethylamino)ethane hydrochloride; N-(2-Chloroethyl)-N,N-dimethylammonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1513422

  SDS
alfa aesar™
A1513422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.05
Chemical Name or Material 2-Dimethylaminoethyl chloride hydrochloride
TSCA TSCA
Assay ≥98%
Molecular Formula ClCH2CH2N(CH3)2·HCl
Melting Point 205° to 208°C
MDL Number MFCD00012516
CAS 4584-46-7
UN Number UN2811
Beilstein 3591234
Quantity 100g
Synonym 1-Chloro-2-(dimethylamino)ethane hydrochloride; N-(2-Chloroethyl)-N,N-dimethylammonium chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number KQ9020000

Applications
2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.

Solubility
Soluble in water (2000 mg/ml at 20°C), and alcohol.

Notes
Hygroscopic, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.