Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dimethylaminobenzoic acid, 98%

$81.12 - $298.62

Chemical Identifiers

Chemical Name or Material 2-Dimethylaminobenzoic acid
CAS 610-16-2
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00016496
Beilstein 2089466
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1296703

  SDS
alfa aesar™
A1296703
1g Each for $81.12

AAA1296706

  SDS
alfa aesar™
A1296706
5g Each for $298.62
Description & Specifications

Specifications

Formula Weight 165.19
Chemical Name or Material 2-Dimethylaminobenzoic acid
TSCA TSCA
Assay 98%
Molecular Formula C9H11NO2
Melting Point 70° to 74°C
MDL Number MFCD00016496
CAS 610-16-2
Beilstein 2089466
Quantity 1g
Odor Odorless

Applications
2-Dimethylaminobenzoic acid is used as a chemical and organic intermediate.

Solubility
Insoluble in water.

Notes
Store in tightly sealed containers in a cool place. Protect from moisture. Incompatible with strong oxidizing agents.