missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1005-28-3 |
---|---|
Molecular Formula | C6H8N2OS |
Molecular Weight (g/mol) | 156.21 |
InChI Key | ZOPJEHPVWLTDJW-UHFFFAOYSA-N |
Synonym | 2-dimethylamino-1,3-thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carboxaldehyde, 2-dimethylamino-thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde,2-dimethylamino, 5-thiazolecarboxaldehyde, 2-dimethylamino, 2-dimethylaminothiazole-5-carbaldehyde |
PubChem CID | 12846795 |
IUPAC Name | 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde |
SMILES | CN(C)C1=NC=C(S1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC444040010
|
Thermo Scientific Chemicals
444040010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1005-28-3 | |
156.21 | |
2-dimethylamino-1,3-thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carboxaldehyde, 2-dimethylamino-thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde,2-dimethylamino, 5-thiazolecarboxaldehyde, 2-dimethylamino, 2-dimethylaminothiazole-5-carbaldehyde | |
2-(dimethylamino)-1,3-thiazole-5-carbaldehyde |
C6H8N2OS | |
ZOPJEHPVWLTDJW-UHFFFAOYSA-N | |
12846795 | |
CN(C)C1=NC=C(S1)C=O |
Specifications
1005-28-3 | |
Glass Bottle | |
1g | |
ZOPJEHPVWLTDJW-UHFFFAOYSA-N | |
2-(dimethylamino)-1,3-thiazole-5-carbaldehyde | |
12846795 | |
97% |
96% min. (GC) | |
C6H8N2OS | |
2-dimethylamino-1,3-thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carboxaldehyde, 2-dimethylamino-thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde,2-dimethylamino, 5-thiazolecarboxaldehyde, 2-dimethylamino, 2-dimethylaminothiazole-5-carbaldehyde | |
CN(C)C1=NC=C(S1)C=O | |
156.21 | |
156.21 | |
2-(Dimethylamino)thiazole-5-carboxaldehyde |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only