Please login to your online account to display your discounted pricing

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H12N2O
Formula Weight 200.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07114CB

  SDS
fisher scientific company
MO07114CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.24
Quantity 250mg
Assay 97%
Molecular Formula C12H12N2O
Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Assay Percent Range 97%
CAS 728035-61-8