Please login to your online account to display your discounted pricing

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO07114CB

 View more versions of this product

Catalog No. MO07114CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.24
Quantity 250mg
Assay 97%
Molecular Formula C12H12N2O
Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Assay Percent Range 97%
CAS 728035-61-8

Specifications

Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H12N2O
Formula Weight 200.24