Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Dimethyl[4-(4-morpholinyl)phenyl]silyl)benzyl alcohol, 95%

$203.57 - $221.68

Chemical Identifiers

Chemical Name or Material 2-(Dimethyl[4-(4-morpholinyl)phenyl]silyl)benzyl alcohol
CAS 1217863-17-6
Molecular Formula C19H25NO2Si
Formula Weight 327.49g/mol
MDL Number MFCD16251558
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5175603

 
alfa aesar™
H51756-03
1g Each for $221.68

AAH5175606

 
alfa aesar™
H51756-06
5g Each for $203.57
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 2-(Dimethyl[4-(4-morpholinyl)phenyl]silyl)benzyl alcohol
CAS 1217863-17-6
Assay 95%
Molecular Formula C19H25NO2Si
Formula Weight 327.49g/mol
MDL Number MFCD16251558
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.