missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF

$96.69 - $370.11

Chemical Identifiers

CAS 1053657-14-9
Molecular Formula C8H20NP
Molecular Weight (g/mol) 161.229
MDL Number MFCD11044863
InChI Key QUVUVMRUSAKOPB-UHFFFAOYSA-N
Synonym 2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity
PubChem CID 57552996
IUPAC Name 2-di(propan-2-yl)phosphanylethanamine
SMILES CC(C)P(CCN)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6043506 View Documents Alfa Aesar™
H6043506
5g Each for $96.69
 
AAH6043514 View Documents Alfa Aesar™
H6043514
25g Each for $370.11
 
Specifications

Chemical Identifiers

1053657-14-9
161.229
QUVUVMRUSAKOPB-UHFFFAOYSA-N
57552996
CC(C)P(CCN)C(C)C
C8H20NP
MFCD11044863
2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity
2-di(propan-2-yl)phosphanylethanamine

Specifications

5g
C8H20NP
MFCD11044863
2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity
QUVUVMRUSAKOPB-UHFFFAOYSA-N
2-di(propan-2-yl)phosphanylethanamine
57552996
2-(Diisopropylphosphino)ethylamine
1053657-14-9
[(CH3)2CH]2PCH2CH2NH2
UN2924
10% w/w soln. in THF
CC(C)P(CCN)C(C)C
161.229
161.22
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.