Learn More
2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6043514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(Diisopropylphosphino)ethylamine | |
C8H20NP | |
MFCD11044863 | |
UN2924 | |
2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity | |
CC(C)P(CCN)C(C)C | |
161.23 | |
161.22 |
1053657-14-9 | |
[(CH3)2CH]2PCH2CH2NH2 | |
25 g | |
Air Sensitive | |
QUVUVMRUSAKOPB-UHFFFAOYSA-N | |
2-di(propan-2-yl)phosphanylethanamine | |
57552996 | |
10% w/w soln. in THF |
Chemical Identifiers
1053657-14-9 | |
161.23 | |
QUVUVMRUSAKOPB-UHFFFAOYSA-N | |
57552996 | |
CC(C)P(CCN)C(C)C |
C8H20NP | |
MFCD11044863 | |
2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity | |
2-di(propan-2-yl)phosphanylethanamine |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335-H336-H351
DOTInformation : Hazard Class: 3; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only