Please login to your online account to display your discounted pricing

2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene
Assay Percent Range 97%
Molecular Formula C7H4F4
Formula Weight 164.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC451370010

 
thermo fisher scientific
AC451370010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC451370050

 
thermo fisher scientific
AC451370050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.1
Quantity 1g
Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene
Assay Percent Range 97%
Molecular Formula C7H4F4
CAS 195886-79-4