Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Difluoromethoxy)benzylamine, 97%

Chemical Identifiers

Chemical Name or Material 2-(Difluoromethoxy)benzylamine
CAS 243863-36-7
Molecular Formula C8H9F2NO
Formula Weight 173.16
MDL Number MFCD00236229
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1916503

  SDS
alfa aesar™
L1916503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19165MD

  SDS
alfa aesar™
L19165MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.16
Chemical Name or Material 2-(Difluoromethoxy)benzylamine
CAS 243863-36-7
MDL Number MFCD00236229
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C8H9F2NO
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
UN Number UN2735

Applications
2-(Difluoromethoxy)benzylamine is used as a pharmaceutical intermediate. It is also used in chemical research.

Notes
Incompatible with oxidizing agents, water and bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.