Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Difluoromethoxy)benzaldehyde, 97%

$62.96 - $212.42

Chemical Identifiers

Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde
CAS 71653-64-0
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00042251
Beilstein 389068
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2427203

 
alfa aesar™
B2427203
1g Each for $62.96

AAB2427206

 
alfa aesar™
B2427206
5g Each for $212.42
Description & Specifications

Specifications

Formula Weight 172.13
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C8H6F2O2
MDL Number MFCD00042251
CAS 71653-64-0
Beilstein 389068
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 106°C (223°F)
Boiling Point 88° to 90°C (9mmHg)
Quantity 1g

Applications
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.

Solubility
Miscible with 2-propanol and acetic acid.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.