Learn More
2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114030050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Diethylaminoethylchloride hydrochloride | |
99.4 | |
208.0°C to 212.0°C | |
1% max. (105°C, 3 hrs) | |
99.5% | |
C6H15Cl2N | |
MFCD00012519 | |
04, III, 240 | |
Solubility in water: 2000g/L (20°C) | |
[H+].[Cl-].CCN(CC)CCCl | |
172.09 | |
172.1 | |
Crystalline Powder |
869-24-9 | |
100.0 | |
White to Beige | |
Authentic | |
Glass Bottle | |
(C2H5)2NCH2CH2Cl·HCl | |
5 g | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
2-chloro-N,N-diethylethanamine;hydrochloride | |
13363 | |
99.5% |
Chemical Identifiers
869-24-9 | |
172.09 | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
13363 | |
[H+].[Cl-].CCN(CC)CCCl |
C6H15Cl2N | |
MFCD00012519 | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
2-chloro-N,N-diethylethanamine;hydrochloride |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 212-786-4
RUO – Research Use Only