Please login to your online account to display your discounted pricing

2-(Diethylamino)-N-(2,6-dimethylphenyl)-acetamide, 97.5%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Diethylamino)-N-(2, 6-dimethylphenyl)-acetamide
CAS 137-58-6
CAS Min % 96.5
CAS Max % 100.0
Assay ≥96.5%
Assay Percent Range 97.5%
Molecular Formula C14H22N2O
Formula Weight 234.34
MDL Number MFCD00026733
Synonym Lidocaine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC137251000

  SDS
thermo fisher scientific
AC137251000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC137250100

  SDS
fisher scientific acros
137250100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.5%
Formula Weight 234.34
Physical Form Crystalline Powder
Quantity 100g
Color White to slightly yellow
Melting Point 66° to 69°C
Merck Index 15, 5535
CAS Max % 100.0
Synonym Lidocaine
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97.5%
Molecular Formula C14H22N2O
Chemical Name or Material 2-(Diethylamino)-N-(2, 6-dimethylphenyl)-acetamide
CAS Min % 96.5
CAS 137-58-6
MDL Number MFCD00026733
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.