Please login to your online account to display your discounted pricing

2-(Diethylamino)-N-(2,6-dimethylphenyl)-acetamide, 97.5%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  137250100

 View more versions of this product

Catalog No. AC137250100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.5%
Formula Weight 234.34
Physical Form Crystalline Powder
Quantity 10g
Color White to slightly yellow
Merck Index 15, 5535
Melting Point 66° to 69°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00026733
Molecular Formula C14H22N2O
CAS Min % 96.5
Assay Percent Range 97.5%
Chemical Name or Material 2-(Diethylamino)-N-(2, 6-dimethylphenyl)-acetamide
CAS 137-58-6
Packaging Glass bottle
Synonym Lidocaine
Infrared Spectrum Authentic
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-(Diethylamino)-N-(2, 6-dimethylphenyl)-acetamide
CAS 137-58-6
CAS Min % 96.5
CAS Max % 100.0
Assay ≥96.5%
Assay Percent Range 97.5%
Molecular Formula C14H22N2O
Formula Weight 234.34
MDL Number MFCD00026733
Synonym Lidocaine
Accessories

Warning

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents