Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Diethylamino)ethanol hydrochloride, 98%

$38.18 - $38.18

Chemical Identifiers

Chemical Name or Material 2-(Diethylamino)ethanol hydrochloride
CAS 14426-20-1
Molecular Formula C6H15NO·HCl
Linear Formula (CH3CH2)2NCH2CH2OH·HCl
Formula Weight 153.65
MDL Number MFCD00036374
Synonym N,N-Diethylethanolamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1656918

 
alfa aesar™
A1656918
50g Each for $38.18
Description & Specifications

Specifications

Formula Weight 153.65
Chemical Name or Material 2-(Diethylamino)ethanol hydrochloride
TSCA TSCA
Assay 98%
Molecular Formula C6H15NO·HCl
Linear Formula (CH3CH2)2NCH2CH2OH·HCl
MDL Number MFCD00036374
CAS 14426-20-1
Melting Point 133° to 137°C
Synonym N,N-Diethylethanolamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number KK5553400
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.