Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl, 98%

$94.70 - $1,858.76

Chemical Identifiers

Chemical Name or Material 2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl
CAS 213697-53-1
Molecular Formula C26H36NP
Formula Weight 393.56g/mol
MDL Number MFCD02183572
Synonym DavePhos; 2-Dimethylamino-2'-dicyclohexylphosphinobiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2767803

 
alfa aesar™
H27678-03
1g Each for $94.70

AAH2767806

 
alfa aesar™
H27678-06
5g Each for $442.53

AAH2767814

 
alfa aesar™
H27678-14
25g Each for $1,858.76
Description & Specifications

Specifications

Melting Point 121° to 124°C
Quantity 1g
Chemical Name or Material 2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl
CAS 213697-53-1
Assay 98%
Molecular Formula C26H36NP
Formula Weight 393.56g/mol
MDL Number MFCD02183572
Synonym DavePhos; 2-Dimethylamino-2'-dicyclohexylphosphinobiphenyl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.